Learn More
ALADDIN SCIENTIFIC CORPORATION MES, 100g
Catalog No. 5000082516
Encompass_Preferred
useful pH range 5 5 - 6 7 pKa 6 1 MES is one of the Good s buffers the most extensively used biological buffers MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring It does not form complexes with majority of the metals used in environmental and biological studies It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes product description MES is one of the Good s buffers the most extensively used biological buffers MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring It does not form complexes with majority of the metals used in environmental and biological studies It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes
Catalog No. 50-000-82516 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. M163014100G
useful pH range 5 5 - 6 7 pKa 6 1 MES is one of the Good s buffers the most extensively used biological buffers MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring It does not form complexes with majority of the metals used in environmental and biological studies It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes product description MES is one of the Good s buffers the most extensively used biological buffers MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring It does not form complexes with majority of the metals used in environmental and biological studies It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes