Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mes (2-morpholinoethanesulphonic acid) | 4432-31-9 | 99.85% | 1 ML
SDP

Catalog No. 5000409843
Encompass_Preferred

MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer designed for various biochemistry and molecular biology applications. It functions effectively within a pH range of 5.5-7.0 and is widely recognized as a Good's buffer, suitable for experiments including cell culture, enzyme activity determination, electrophoresis, and protein studies.

  • Amphoteric ion buffer
  • Buffer capacity from pH 5.5-7.0
  • Ideal for cell culture, enzyme activity determination, electrophoresis, and protein studies
  • Solid, white to off-white appearance
  • Soluble in water and DMSO

Catalog No. 50-004-09843 Supplier Medchemexpress LLC Supplier No. HYD085810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer designed for various biochemistry and molecular biology applications. It functions effectively within a pH range of 5.5-7.0 and is widely recognized as a Good's buffer, suitable for experiments including cell culture, enzyme activity determination, electrophoresis, and protein studies.

  • Amphoteric ion buffer
  • Buffer capacity from pH 5.5-7.0
  • Ideal for cell culture, enzyme activity determination, electrophoresis, and protein studies
  • Solid, white to off-white appearance
  • Soluble in water and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.