Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals MES, 99+%, <1% water

Catalog No. AC274140500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC274140500 50 g Glass bottle
AC274142500 250 g Glass bottle
AC274140010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC274140500 Supplier Thermo Scientific Chemicals Supplier No. 274140500
  • $136.10
    $114.65 / Each of 1

    Save $21.45

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4432-31-9 | C6H13NO4S | 195.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
MDL Number MFCD00006181
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic acid
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O

Specifications

Melting Point >300.0°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Packaging Glass bottle
Merck Index 15, 5972
Solubility Information Solubility in water: 10g/100mL.
Formula Weight 195.24
Percent Purity ≥99%
Chemical Name or Material MES
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Use only outdoors or in a well-ventilated area.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.