Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp MES, Anhydrous, 250 Grams
SDP

Catalog No. 501368363
Encompass_Preferred

  • CAS Number: 4432-31-9
  • Chemical Formula: C6H13NO4S
  • Molecular Weight: 195.24 g/mol

Catalog No. 50-136-8363 Supplier Research Products International Corp Supplier No. M22580250.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • CAS Number: 4432-31-9
  • Chemical Formula: C6H13NO4S
  • Molecular Weight: 195.24 g/mol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.