Learn More
Boston Bioproducts Inc MES Buffer (0.5 M, pH 6.0) - 250ml
Catalog No. NC0440019
Encompass
MES Buffer [2- (N-morpholino) ethanesulfonic acid] is a zwitterionic Good's buffer used in biological and biochemical research. It maintains a stable pH within the range of 5.5 to 7.0 and is highly soluble in water. MES is commonly used as a running buffer for bis-tris gels, chromatography, and in protein purification. It is valuable for enzymatic reactions, and acts as a non-coordinating buffer in metal ion chemistry. It has minimal UV absorbance and is stable in various applications.
Catalog No. NC0440019 Supplier Boston Bioproducts Inc Supplier No. BB114250ML
MES Buffer [2- (N-morpholino) ethanesulfonic acid] is a zwitterionic Good's buffer used in biological and biochemical research. It maintains a stable pH within the range of 5.5 to 7.0 and is highly soluble in water. MES is commonly used as a running buffer for bis-tris gels, chromatography, and in protein purification. It is valuable for enzymatic reactions, and acts as a non-coordinating buffer in metal ion chemistry. It has minimal UV absorbance and is stable in various applications.