Learn More
Boston Bioproducts Inc MES Buffer (1 M, pH 5.0) - 250ml
Catalog No. NC1453919
Encompass
MES Buffer [2- (N-morpholino) ethanesulfonic acid] is a zwitterionic Good's buffer used in biological and biochemical research. It maintains a stable pH within the range of 5.5 to 7.0 and is highly soluble in water. MES is commonly used as a running buffer for bis-tris gels, chromatography, and in protein purification. It is valuable for enzymatic reactions, and acts as a non-coordinating buffer in metal ion chemistry. It has minimal UV absorbance and is stable in various applications.
Catalog No. NC1453919 Supplier Boston Bioproducts Inc Supplier No. BBMS50250ML
Add to Cart
Add to Cart
MES Buffer [2- (N-morpholino) ethanesulfonic acid] is a zwitterionic Good's buffer used in biological and biochemical research. It maintains a stable pH within the range of 5.5 to 7.0 and is highly soluble in water. MES is commonly used as a running buffer for bis-tris gels, chromatography, and in protein purification. It is valuable for enzymatic reactions, and acts as a non-coordinating buffer in metal ion chemistry. It has minimal UV absorbance and is stable in various applications.