Learn More
Bioworld MES Buffer 1M, pH 6.1n 4432-31-9, 250ml
Catalog No. NC0288420
Encompass_Preferred
Also known as [2-(N-Morpholino) ethanesulfonic Acid]. MES-Buffer has a pKa value near physiological pH, it is not absorbed through cell membranes, and it is essentially transparent to UV light. A biological buffer mainly used in plant cell cultures. Technical Information: Used for culture media. Ultra-Pure Grade, pKa (at 25°C): 6.10+/-0.05, pH 6.1, Molecular Weight : 195.24, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : >300 °C(lit.) 250ml
Catalog No. NC0288420 Supplier Bioworld Supplier No. 413200361
Also known as [2-(N-Morpholino) ethanesulfonic Acid]. MES-Buffer has a pKa value near physiological pH, it is not absorbed through cell membranes, and it is essentially transparent to UV light. A biological buffer mainly used in plant cell cultures. Technical Information: Used for culture media. Ultra-Pure Grade, pKa (at 25°C): 6.10+/-0.05, pH 6.1, Molecular Weight : 195.24, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : >300 °C(lit.) 250ml