Learn More
Alkali Scientific MES Buffer, Anhydrous, 5 Kg
SDP
Catalog No. 5000660096
Encompass_Preferred
MES (2-(N-morpholino)ethanesulfonic acid) anhydrous is a biochemical reagent designed for pH buffering in laboratory applications. This 5kg volume of MES anhydrous is used to prepare MES buffers that provide a stable pH range, particularly for biological assays and biochemical studies. MES is widely used in enzyme reactions, protein stabilization, and tissue culture media where maintaining a consistent acidic pH is critical. It is particularly effective at buffering in the pH range of 5.5 to 6.7, making it suitable for various biochemical processes, including those in protein chemistry and molecular biology. MES is often utilized in electrophoresis, chromatography, and other sensitive experiments requiring precise pH control.
Catalog No. 50-006-60096 Supplier Alkali Scientific Supplier No. MEA003CP
Add to Cart
Add to Cart
MES (2-(N-morpholino)ethanesulfonic acid) anhydrous is a biochemical reagent designed for pH buffering in laboratory applications. This 5kg volume of MES anhydrous is used to prepare MES buffers that provide a stable pH range, particularly for biological assays and biochemical studies. MES is widely used in enzyme reactions, protein stabilization, and tissue culture media where maintaining a consistent acidic pH is critical. It is particularly effective at buffering in the pH range of 5.5 to 6.7, making it suitable for various biochemical processes, including those in protein chemistry and molecular biology. MES is often utilized in electrophoresis, chromatography, and other sensitive experiments requiring precise pH control.