Learn More
Medchemexpress LLC MES-d13 | 352534-94-2 | 98.8% | 5 MG
SDP
Catalog No. 5000410166
Encompass_Preferred
MES-d13 is the deuterium labeled MES. This amphoteric ion buffer has a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is utilized in various biochemical and molecular biology experiments.
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Applicable in cell culture
  • Useful for enzyme activity determination
  • Used in electrophoresis
  • Applied in protein studies
Catalog No. 50-004-10166 Supplier Medchemexpress LLC Supplier No. HYD0858S5MG
MES-d13 is the deuterium labeled MES. This amphoteric ion buffer has a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is utilized in various biochemical and molecular biology experiments.
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Applicable in cell culture
  • Useful for enzyme activity determination
  • Used in electrophoresis
  • Applied in protein studies