Learn More
Medchemexpress LLC MES-d13 | 352534-94-2 | 98.8% | 50 MG
SDP
Catalog No. 5000409987
Encompass_Preferred
MES-d13 is the deuterium labeled MES. MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, MES is widely used in biochemistry and molecular biology experiments, such as cell culture, enzyme activity determination, electrophoresis, and protein studies. For research use only, not for sale to patients.
  • Deuterium labeled MES
  • Amphoteric ion buffer
  • Buffer capacity ranging from pH 5.5-7.0
  • Used in biochemistry and molecular biology experiments
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies
Catalog No. 50-004-09987 Supplier Medchemexpress LLC Supplier No. HYD0858S50MG
MES-d13 is the deuterium labeled MES. MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, MES is widely used in biochemistry and molecular biology experiments, such as cell culture, enzyme activity determination, electrophoresis, and protein studies. For research use only, not for sale to patients.
  • Deuterium labeled MES
  • Amphoteric ion buffer
  • Buffer capacity ranging from pH 5.5-7.0
  • Used in biochemistry and molecular biology experiments
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies