Learn More
Teknova MES Dilution Buffer, pH 6.5. 1000mL, Sterile 8 pack.
SDP
Catalog No. 502745800
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-274-5800 Supplier Teknova Supplier No. M51908PK
Add to Cart
Add to Cart
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.