Learn More
Teknova MES Elution Buffer, pH 6.5. 1000mL, Sterile
SDP
Catalog No. 502745857
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-274-5857 Supplier Teknova Supplier No. M2280
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.