Learn More
Gold Biotechnology Inc MES Free Acid Monohydrate
SDP
Catalog No. 5000562098
Encompass_Preferred
MES monohydrate is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic morpholinic buffer that is useful for a pH range of 5 5 6 7 and commonly used for cell culture media as a running buffer in electrophoresis and for protein purification in chromatography MES lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions
Catalog No. 50-005-62098 Supplier Gold Biotechnology Inc Supplier No. M090500
MES monohydrate is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic morpholinic buffer that is useful for a pH range of 5 5 6 7 and commonly used for cell culture media as a running buffer in electrophoresis and for protein purification in chromatography MES lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions