Learn More
Gold Biotechnology Inc MES Free Acid Monohydrate, Ultra Pure 100 g
SDP
Catalog No. NC1563255
Encompass_Preferred
MES monohydrate is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al. It is a zwitterionic, morpholinic buffer that is useful for a pH range of 5.5 – 6.7 and commonly used for cell culture media, as a running buffer in electrophoresis, and for protein purification in chromatography. MES lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions. MES is often used in buffered culture media for bacteria, yeast, and mammalian cells. It is toxic towards plants at high concentrations, but can be used in culture media at lower concentrations for plants. Due to low ionic mobility, MES is regarded as an excellent buffer for use in capillary electrochromatography.
Catalog No. NC1563255 Supplier Gold Biotechnology Inc Supplier No. M090100
Add to Cart
Add to Cart
MES monohydrate is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al. It is a zwitterionic, morpholinic buffer that is useful for a pH range of 5.5 – 6.7 and commonly used for cell culture media, as a running buffer in electrophoresis, and for protein purification in chromatography. MES lacks the ability to form a complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions. MES is often used in buffered culture media for bacteria, yeast, and mammalian cells. It is toxic towards plants at high concentrations, but can be used in culture media at lower concentrations for plants. Due to low ionic mobility, MES is regarded as an excellent buffer for use in capillary electrochromatography.