Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MES hydrate | 1266615-59-1 | 99.9% | 5 G
SDP

Catalog No. 5000410060
Encompass_Preferred

MES (2-Morpholinoethanesulphonic acid) hydrate is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. It is a zwitterionic buffer that provides effective buffering capacity around pH 6.15. Its pKa value changes with temperature. MES hydrate exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.

  • Appearance: solid
  • Color: white to off-white
  • Formula: C6H13NO4S.xH2O
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: store at room temperature, keep dry and cool. In solvent: -80°C for 1 year, -20°C for 6 months
  • Solubility: H2O : 175 mg/mL (requires ultrasonic)

Catalog No. 50-004-10060 Supplier Medchemexpress LLC Supplier No. HYD0858C5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MES (2-Morpholinoethanesulphonic acid) hydrate is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. It is a zwitterionic buffer that provides effective buffering capacity around pH 6.15. Its pKa value changes with temperature. MES hydrate exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.

  • Appearance: solid
  • Color: white to off-white
  • Formula: C6H13NO4S.xH2O
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: store at room temperature, keep dry and cool. In solvent: -80°C for 1 year, -20°C for 6 months
  • Solubility: H2O : 175 mg/mL (requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.