Learn More
Medchemexpress LLC MES hydrate | 1266615-59-1 | MFCD00149409 | 99.9% | 10 G
SDP
Catalog No. 5000409841
Encompass_Preferred
MES (2-Morpholinoethanesulphonic acid) hydrate is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. It is classified as a Good's buffer and is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. This zwitterionic buffer provides effective buffering capacity around pH 6.15. Its pKa value changes with temperature, a factor to consider when used at varying temperatures. MES hydrate exhibits good buffering properties in biological experiments and has a poor affinity for metal ions, making it suitable for studies involving such ions.
  • Amphoteric ion buffer
  • Buffer capacity ranging pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Effective buffering capacity at approximately pH 6.15
  • Poor affinity with metal ions
Catalog No. 50-004-09841 Supplier Medchemexpress LLC Supplier No. HYD0858C10G
MES (2-Morpholinoethanesulphonic acid) hydrate is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. It is classified as a Good's buffer and is widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. This zwitterionic buffer provides effective buffering capacity around pH 6.15. Its pKa value changes with temperature, a factor to consider when used at varying temperatures. MES hydrate exhibits good buffering properties in biological experiments and has a poor affinity for metal ions, making it suitable for studies involving such ions.
  • Amphoteric ion buffer
  • Buffer capacity ranging pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Effective buffering capacity at approximately pH 6.15
  • Poor affinity with metal ions