Learn More
Sigma Aldrich Fine Chemicals Biosciences MES hydrate 99.5 titration50KG
Catalog No. 502914453
Encompass
MES is one of the Good buffers the most extensively used biological buffers. MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring. It does not form complexes with majority of the metals used in environmental and biological studies. It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes.
Catalog No. 50-291-4453 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M825050KG
Add to Cart
Add to Cart
Request a Quote
MES is one of the Good buffers the most extensively used biological buffers. MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring. It does not form complexes with majority of the metals used in environmental and biological studies. It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes.