Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals MES hydrate, 99+% PROMO

Catalog No. H5647236F25
Change view
Click to view available options
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
H5647236F25 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H5647236F25 Supplier Thermo Scientific Chemicals Supplier No. H5647236PR/DEL

missing translation for 'validMainframeMsg'

MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications.

  • Applications: A useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
  • Solubility: Soluble in water. Slightly soluble in organic solvents.
  • Notes: Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23 g/mol
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point 308 °C (Decomposition)
Quantity 500 g
Beilstein 141319
Merck Index 14,5902
Solubility Information Soluble in Water. Slightly Soluble in Organic Solvents.
Formula Weight 195.24 amu (Anhydrous)
Percent Purity ≥99%
Chemical Name or Material MES hydrate
Hazard Category Warning
Hazard Statement

GHS H Statement

  • H315-H319-H335
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
Precautionary Statement

GHS P Statement

  • P261-P280-P305+P351+P338-P304+P340-P405-P501a
  • Avoid breathing dust/fume/gas/mist/vapors/spray.
  • Wear protective gloves/protective clothing/eye protection/face protection.
  • IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing
  • Store locked up.
  • Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 224-632-3
RTECSNumber QE3479500
TSCA Yes
Recommended Storage Ambient Temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.