Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES, Monohydrate, cGMP Manufactured, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18604327 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
12 kg
25 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-327 25 kg Poly Bottle
18-604-325 1 kg Poly Bottle
18-604-326 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-327 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M284825KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M2848, 145224-94-8, C6H13NO4S·H2O

Spectrum™ Chemical MES, Monohydrate, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

CAS Max % 100%
pH 2.5 to 4.0
Physical Form Powder
Quantity 25 kg
MDL Number MFCD00149409
Packaging Poly Bottle
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material 2-(N-Morpholino)Ethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.