Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES monohydrate, free acid, ≥99%, MP Biomedicals™

Catalog No. ICN19483580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19483580 100 g
ICN19483510 10 g
ICN19483583 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19483580 Supplier MP Biomedicals, Inc Supplier No. 0219483580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa ± 1). Alternatively, volumes of equimolar MES free acid and sodium or potassium MES can be mixed to attain the desired pH

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-[N-morpholino]ethanesulfonic acid
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Solution Type Buffer
Assay Percent Range ≥99%
Formula Weight 213.26
Percent Purity 99.6%
Grade Molecular Biology
Chemical Name or Material MES monohydrate
EINECSNumber 224-632-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.