Learn More
Medchemexpress LLC MES monohydrate (Standard) | 145224-94-8 | MFCD00149409 | 100.0% | 25 MG
SDP
Catalog No. 5000412548
Encompass_Preferred
MES monohydrate (Standard) is an analytical standard and an amphoteric ion buffer used in research and analytical applications. This compound, also known as 2-Morpholinoethanesulphonic acid, is a Good's buffer with a capacity ranging from pH 5.5-7.0.
  • Analytical standard for research and analytical applications
  • Functions as an amphoteric ion buffer
  • Offers a buffer capacity from pH 5.5-7.0
  • Widely used in biochemistry and molecular biology
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-004-12548 Supplier Medchemexpress LLC Supplier No. 25MGHYD0858AR
MES monohydrate (Standard) is an analytical standard and an amphoteric ion buffer used in research and analytical applications. This compound, also known as 2-Morpholinoethanesulphonic acid, is a Good's buffer with a capacity ranging from pH 5.5-7.0.
  • Analytical standard for research and analytical applications
  • Functions as an amphoteric ion buffer
  • Offers a buffer capacity from pH 5.5-7.0
  • Widely used in biochemistry and molecular biology
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS