Learn More
MP Biomedicals™ MES SDS Running Buffer for Bis-Tris Gels, Powder
Shop All MP Biomedicals, Inc ProductsDescription
MES SDS Running Buffer (4-morpholinoethanesulfonic acid) for Bis-Tris Gels in Powder (One Pack makes up 1 L)
Specifications
Specifications
| Concentration | 1X |
| For Use With (Application) | Protein Electrophoresis |
| For Use With (Equipment) | SDS-Page |
| Physical Form | Powder |
| Quantity | 5 Pouches |
| Solution Type | Buffer |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.