Learn More
Sigma Aldrich Fine Chemicals Biosciences MES SDS running buffer pow
Catalog No. 502901738
Encompass
MES SDS running buffer powder for mPAGEBis-Tris gels. 5 packets. Each paclet makes 1 liter of running buffer.Unless otherwise stated in our catalog or other company documentation accompanying the product(s) our products are intended for research use only and are not to be used for any other purpose which includes but is not limited to unauthorized commercial uses in vitro diagnostic uses ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Catalog No. 50-290-1738 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MPMES
MES SDS running buffer powder for mPAGEBis-Tris gels. 5 packets. Each paclet makes 1 liter of running buffer.Unless otherwise stated in our catalog or other company documentation accompanying the product(s) our products are intended for research use only and are not to be used for any other purpose which includes but is not limited to unauthorized commercial uses in vitro diagnostic uses ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.