Learn More
Alkali Scientific MES Sodium Salt [2 (N Morpholino)ethanesulfonic acid] sodium salt, 5 Kg
SDP
Catalog No. 5000659869
Encompass_Preferred
MES Sodium Salt [2-(N-Morpholino)ethanesulfonic acid] Sodium Salt, 5 Kg, is a widely used buffering reagent known for its ability to stabilize pH in a variety of biological and biochemical applications. With a high buffering capacity, MES is effective in maintaining pH in the range of 5.5 to 6.7, making it ideal for enzyme reactions, electrophoresis, and other experiments requiring precise pH control. The 5 kg weight offers a cost-effective solution for large-scale operations or frequent use. MES is often used in protein chemistry, molecular biology, and cell culture media to ensure consistent conditions during experimental procedures.
Catalog No. 50-006-59869 Supplier Alkali Scientific Supplier No. ESS004CP
Add to Cart
Add to Cart
MES Sodium Salt [2-(N-Morpholino)ethanesulfonic acid] Sodium Salt, 5 Kg, is a widely used buffering reagent known for its ability to stabilize pH in a variety of biological and biochemical applications. With a high buffering capacity, MES is effective in maintaining pH in the range of 5.5 to 6.7, making it ideal for enzyme reactions, electrophoresis, and other experiments requiring precise pH control. The 5 kg weight offers a cost-effective solution for large-scale operations or frequent use. MES is often used in protein chemistry, molecular biology, and cell culture media to ensure consistent conditions during experimental procedures.