Learn More
Sigma Aldrich Fine Chemicals Biosciences MES sodium salt 99 titrati25G
Catalog No. 502899883
Encompass
MES sodium salt is commonly employed as buffer. Useful pH range for MES sodium salt is 5.5 to 6.7.
Catalog No. 50-289-9883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M388525G
MES sodium salt is commonly employed as buffer. Useful pH range for MES sodium salt is 5.5 to 6.7.