Learn More
Sigma Aldrich Fine Chemicals Biosciences MES sodium salt BioPerform100G
Catalog No. 502891494
Encompass
MES is one of the so-called Good buffers developed for biological applications. This product is BioPerformance certified (suitable for electrophoresis, tissue culture, and molecular biology applications. A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa/-1). Alternatively, volumes of equimolar solutions of MES free acid and MES sodium can be mixed to attain the desired pH. Standard mixing tables using stock solutions to prepare a buffer of a given pH have been published.
Catalog No. 50-289-1494 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M3058100G
MES is one of the so-called Good buffers developed for biological applications. This product is BioPerformance certified (suitable for electrophoresis, tissue culture, and molecular biology applications. A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa/-1). Alternatively, volumes of equimolar solutions of MES free acid and MES sodium can be mixed to attain the desired pH. Standard mixing tables using stock solutions to prepare a buffer of a given pH have been published.