Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MES, ULTROL™ Grade, Calbiochem™,

Catalog No. 4758931KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-589-31KG 1 kg
47-589-3100GM 100 g
47-589-3500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 47-589-31KG Supplier MilliporeSigma™ Supplier No. 4758931KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A zwitterionic buffer useful in the pH range of 5.5 to 6.5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic Acid
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O

Specifications

Quantity 1 kg
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 195.2
Grade by titration (dry basis)
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.