Learn More
Selleck Chemical LLC Mesalamine 5-ASA-50mg
SDP
Catalog No. 502389929
Encompass_Preferred
Mesalazine (5-ASA) is a specific inhibitor of TNFα-induced IKK activity, used to treat inflammatory bowel disease.
Catalog No. 50-238-9929 Supplier Selleck Chemical LLC Supplier No. S168150MG
Mesalazine (5-ASA) is a specific inhibitor of TNFα-induced IKK activity, used to treat inflammatory bowel disease.