Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesamoll™ Selectophore, MilliporeSigma™ Supelco™

Catalog No. 111015890 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5890 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5890 Supplier MilliporeSigma Supelco Supplier No. 509875ML
Add to Cart
Add to Cart

Mesamoll™ is an alkyl sulfonate plasticizer. Owing to its alkali resistance nature, it is widely used in membranes and sealants in the construction industry.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91082-17-6
Product Line Mesamoll
Flash Point 210°C to 240°C
Synonym Alkylsulfonic phenyl ester
Type Organic Standard
Quantity 5 mL
Grade Selectophore

Mesamoll is a registered trademark of Lanxess Deutschland GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.