Learn More
Lonza Walkersville MsGM™ Mesangial Cell Growth Medium BulletKit™
Catalog No. NC2953765
Encompass
Culture system containing MsGMTM Mesangial Basal Medium (CC-3147) and MsGMTM SingleQuotsTM Supplements (CC-4146) required for growth of human mesangial cells.
Catalog No. NC2953765 Supplier Lonza Walkersville Supplier No. CC3146
Culture system containing MsGMTM Mesangial Basal Medium (CC-3147) and MsGMTM SingleQuotsTM Supplements (CC-4146) required for growth of human mesangial cells.