Learn More
Sciencell Research Laboratories MESANGIAL CELL MEDIUM 500 ML
Catalog No. NC9984905
Encompass
Mesangial Cell medium is a complete medium designed for optimal growth of normal human renal mesangial cells in vitro. It is shipped as a kit with mesangial cell growth supplement and Pen/Strep solution.
Catalog No. NC9984905 Supplier Sciencell Research Laboratories Supplier No. 4201
Mesangial Cell medium is a complete medium designed for optimal growth of normal human renal mesangial cells in vitro. It is shipped as a kit with mesangial cell growth supplement and Pen/Strep solution.