Learn More
Medchemexpress LLC Mesdopetam hemitartrate | 2562346-14-7 | 99.86% | 350.39 | 50 MG
SDP
Catalog No. 5000344255
Encompass_Preferred
Mesdopetam (IRL790) hemitartrate is a dopamine D3 receptor antagonist with psychomotor stabilizing properties, exhibiting a Ki of 90 nM and an IC50 of 9.8 μM for the human recombinant D3 receptor. It is utilized in research for motor and psychiatric complications associated with Parkinson disease.
  • Dopamine D3 receptor antagonist
  • Psychomotor stabilizing properties
  • Used for research of motor and psychiatric complications in Parkinson disease
  • Exhibits a Purity of 99.86%
Catalog No. 50-003-44255 Supplier Medchemexpress LLC Supplier No. HY109150A100MG
Mesdopetam (IRL790) hemitartrate is a dopamine D3 receptor antagonist with psychomotor stabilizing properties, exhibiting a Ki of 90 nM and an IC50 of 9.8 μM for the human recombinant D3 receptor. It is utilized in research for motor and psychiatric complications associated with Parkinson disease.
  • Dopamine D3 receptor antagonist
  • Psychomotor stabilizing properties
  • Used for research of motor and psychiatric complications in Parkinson disease
  • Exhibits a Purity of 99.86%