Learn More
Medchemexpress LLC Mesdopetam hemitartrate | 2562346-14-7 | 99.9% | 350.39 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY109150A10MG
Encompass_Preferred
Mesdopetam (IRL790) hemitartrate is a dopamine D3 receptor antagonist (Ki=90 nM; IC50=9.8 μM for human recombinant D3 receptor) with psychomotor stabilizing properties. Mesdopetam hemitartrate is used for the research of motor and psychiatric complications in Parkinson disease.
  • Dopamine D3 receptor antagonist (Ki=90 nM; IC50=9.8 μM for human recombinant D3 receptor).
  • Possesses psychomotor stabilizing properties.
  • Utilized for the research of motor and psychiatric complications in Parkinson disease.
  • Inhibits behavioral activation dose-dependently following pretreatment with D-amphetamine or MK-801.
Catalog No. 50-003-46534