Learn More
Medchemexpress LLC Mesdopetam hemitartrate | 2562346-14-7 | 99.9% | 350.39 | 50 MG
SDP
Catalog No. 5000343375
Encompass_Preferred
Mesdopetam hemitartrate is a dopamine D3 receptor antagonist with psychomotor stabilizing properties. It is used for the research of motor and psychiatric complications in Parkinson disease.
  • Dopamine D3 receptor antagonist
  • Psychomotor stabilizing properties
  • Used for research of motor and psychiatric complications in Parkinson disease
Catalog No. 50-003-43375 Supplier Medchemexpress LLC Supplier No. HY109150A50MG
Mesdopetam hemitartrate is a dopamine D3 receptor antagonist with psychomotor stabilizing properties. It is used for the research of motor and psychiatric complications in Parkinson disease.
  • Dopamine D3 receptor antagonist
  • Psychomotor stabilizing properties
  • Used for research of motor and psychiatric complications in Parkinson disease