Learn More
Medchemexpress LLC Mesdopetam hemitartrate | 2562346-14-7 | 99.9% | 350.39 | C12H18FNO3S·1/2C4H6O6 | 5 MG
SDP
Catalog No. 5000264439
Encompass_Preferred
Mesdopetam hemitartrate is the hemitartrate salt of mesdopetam (IRL790), a dopamine D3-preferring receptor antagonist supplied for research use. It is intended for in vitro and in vivo studies of motor and psychiatric complications associated with Parkinson's disease and is supplied as a white to off-white solid with high purity.
  • Dopamine D3 receptor antagonist activity suitable for neuroscience research.
  • High purity (≈99.9%) for reliable experimental results.
  • Available in small solid quantities and as a DMSO solution for convenience.
  • Stable when stored sealed and protected from moisture under recommended temperatures.
  • Downloadable datasheet and SDS provided for experimental and safety information.
Catalog No. 50-002-64439 Supplier Medchemexpress LLC Supplier No. HY109150A5MG
Mesdopetam hemitartrate is the hemitartrate salt of mesdopetam (IRL790), a dopamine D3-preferring receptor antagonist supplied for research use. It is intended for in vitro and in vivo studies of motor and psychiatric complications associated with Parkinson's disease and is supplied as a white to off-white solid with high purity.
  • Dopamine D3 receptor antagonist activity suitable for neuroscience research.
  • High purity (≈99.9%) for reliable experimental results.
  • Available in small solid quantities and as a DMSO solution for convenience.
  • Stable when stored sealed and protected from moisture under recommended temperatures.
  • Downloadable datasheet and SDS provided for experimental and safety information.