Learn More
STEMCELL Technologies STEMdiff™ Mesenchymal Progenitor Kit, Size: 1 Kit
SDP
Catalog No. NC1382645
Encompass
STEMdiff™ Mesenchymal Progenitor Kit is a defined culture kit consisting of animal component-free (ACF) induction medium, expansion medium, and attachment substrate. It is optimized for the derivation of cells with mesenchymal progenitor cell (MPC)-like properties from human embryonic stem (ES) cells or induced pluripotent stem (iPS) cells. This kit provides a complete workflow of defined reagents for derivation and expansion of human ES- or iPS-derived MPCs.CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.
Catalog No. NC1382645 Supplier STEMCELL Technologies Supplier No. 05240
STEMdiff™ Mesenchymal Progenitor Kit is a defined culture kit consisting of animal component-free (ACF) induction medium, expansion medium, and attachment substrate. It is optimized for the derivation of cells with mesenchymal progenitor cell (MPC)-like properties from human embryonic stem (ES) cells or induced pluripotent stem (iPS) cells. This kit provides a complete workflow of defined reagents for derivation and expansion of human ES- or iPS-derived MPCs.CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.