Learn More
ATCC Mesenchymal Stem Cell Basal Medium, 485mL
Catalog No. 502383945
Encompass
Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical, and Bone Marrow-derived MSCs is a sterile, phenol red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This low serum (2% FBS) system supports mesenchymal stem cells derived from various normal human tissues lipoaspirates, and umbilical cord. Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical, and Bone Marrow-derived MSCs contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. To support the proliferation and plating efficiency of various adult stem cells, Mesenchymal Stem Cell Basal Medium with the appropriate cell-specific growth kit. When using this complete media system, the growth of undifferentiated mesenchymal stem cells does not need feeder layers, extracellular matrix proteins, or other substrates.
Catalog No. 50-238-3945 Supplier ATCC Supplier No. PCS500030
Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical, and Bone Marrow-derived MSCs is a sterile, phenol red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This low serum (2% FBS) system supports mesenchymal stem cells derived from various normal human tissues lipoaspirates, and umbilical cord. Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical, and Bone Marrow-derived MSCs contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. To support the proliferation and plating efficiency of various adult stem cells, Mesenchymal Stem Cell Basal Medium with the appropriate cell-specific growth kit. When using this complete media system, the growth of undifferentiated mesenchymal stem cells does not need feeder layers, extracellular matrix proteins, or other substrates.