Learn More
ATCC Mesenchymal Stem Cell Growth Kit-Low Serum for Adipose and Umbilical Cord-derived MSCs
Catalog No. 502381751
Encompass
Mesenchymal Stem Cell Growth Kit for Adipose and Umbilical-derived MSCs - Low Serum contains components that when added to Mesenchymal Stem Cell Basal Medium (ATCC PCS-500-030) create a complete ATCC Primary Cell Solutions culture environment for multipotent stem cells derived from normal human tissues including lipoaspirates (eg, Adipose-derived Mesenchymal Stem Cells; Normal, Human, ATCC PCS-500-011) or umbilical cord (Umbilical Cord-derived Mesenchymal Stem Cells; Normal, Human, ATCC PCS-500-010). The low serum (2% FBS) medium formulation is designed to support normal, undifferentiated mesenchymal stem cell morphology and promotes rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-1751 Supplier ATCC Supplier No. PCS500040
Add to Cart
Add to Cart
Mesenchymal Stem Cell Growth Kit for Adipose and Umbilical-derived MSCs - Low Serum contains components that when added to Mesenchymal Stem Cell Basal Medium (ATCC PCS-500-030) create a complete ATCC Primary Cell Solutions culture environment for multipotent stem cells derived from normal human tissues including lipoaspirates (eg, Adipose-derived Mesenchymal Stem Cells; Normal, Human, ATCC PCS-500-011) or umbilical cord (Umbilical Cord-derived Mesenchymal Stem Cells; Normal, Human, ATCC PCS-500-010). The low serum (2% FBS) medium formulation is designed to support normal, undifferentiated mesenchymal stem cell morphology and promotes rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.