Learn More
ATCC Mesenchymal Stem Cell Growth Kit for Bone Marrow-derived MSCs
Catalog No. 502382361
Encompass
Mesenchymal Stem Cell Growth Kit for Bone Marrow-derived MSCs contains components that when added to Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical and Bone Marrow-derived MSCs create a complete culture environment for multipotent stem cells derived from normal human bone marrow (eg, Bone Marrow-derived MSCs, ATCC PCS-500-012). The serum (7% FBS) medium formulation is designed to support normal, undifferentiated mesenchymal stem cell morphology and promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-2361 Supplier ATCC Supplier No. PCS500041
Mesenchymal Stem Cell Growth Kit for Bone Marrow-derived MSCs contains components that when added to Mesenchymal Stem Cell Basal Medium for Adipose, Umbilical and Bone Marrow-derived MSCs create a complete culture environment for multipotent stem cells derived from normal human bone marrow (eg, Bone Marrow-derived MSCs, ATCC PCS-500-012). The serum (7% FBS) medium formulation is designed to support normal, undifferentiated mesenchymal stem cell morphology and promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.