Learn More
FUJIFILM Biosciences mesenchymal stem cell XSFM 1L
Catalog No. 502992660
Encompass_Preferred
PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expansion of purified or enriched human mesenchymal stromal cells (MSCs) under serum-free and xeno-free culture conditions. This medium is ready to use. It may also be used with additional cytokine/growth factors for desired application.
Catalog No. 50-299-2660 Supplier FUJIFILM Biosciences Supplier No. 911491L
PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expansion of purified or enriched human mesenchymal stromal cells (MSCs) under serum-free and xeno-free culture conditions. This medium is ready to use. It may also be used with additional cytokine/growth factors for desired application.