Learn More
ATCC Mesenchymal Stem Cells; derived from BYS0112; Induced Pluripotent Stem Cell; Human (Homo sapiens)
Supplier:  ATCC ACS7010
Encompass
iPSC-derived Mesenchymal Stem Cells; BYS0112 are induced pluripotent stem cells that have been differentiated into multipotent stromal cells. iPSC-derived MSCs may be further differentiated into osteoblasts, chondrocytes, myocytes, and adipocytes. These cells can be used in bone cell lineage differentiation, regenerative medicine, cell therapy, exosome research, and cancer immunology research.
Catalog No. 50-238-2566