Learn More
STEMCELL Technologies MesenCult™ Adipogenic Differentiation Kit (Human), Size: 250 mL
SDP
Catalog No. NC0690389
Encompass
MesenCult™ Adipogenic Differentiation Medium (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into adipogenic lineage cells. This kit is suitable for the differentiation of MSCs derived from human bone marrow (BM), adipose tissue, umbilical cord, or pluripotent stem cells (PSCs) that have been previously culture-expanded in serum- and animal component-free medium (e.g. MesenCult™ ACF Plus Medium [Catalog #05445]), serum-containing medium (e.g. MesenCult™ Proliferation Kit [Catalog #05411]), or platelet lysate medium (e.g. MesenCult™-hPL Medium [Catalog #05439]).
Catalog No. NC0690389 Supplier STEMCELL Technologies Supplier No. 05412
MesenCult™ Adipogenic Differentiation Medium (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into adipogenic lineage cells. This kit is suitable for the differentiation of MSCs derived from human bone marrow (BM), adipose tissue, umbilical cord, or pluripotent stem cells (PSCs) that have been previously culture-expanded in serum- and animal component-free medium (e.g. MesenCult™ ACF Plus Medium [Catalog #05445]), serum-containing medium (e.g. MesenCult™ Proliferation Kit [Catalog #05411]), or platelet lysate medium (e.g. MesenCult™-hPL Medium [Catalog #05439]).