Learn More
STEMCELL Technologies MesenCult™ MSC Basal Medium (Human), Size: 450 mL
SDP
Catalog No. 501974344
Encompass
MesenCult™ MSC Basal Medium (Human) is a standardized basal medium designed to be supplemented with MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human; Catalog #05402) for the in vitro culture of human mesenchymal stem cells (MSCs). MesenCult™ MSC Basal Medium is a component of MesenCult™ Proliferation Kit (Human; Catalog #05411), and is also available separately.
Catalog No. 50-197-4344 Supplier STEMCELL Technologies Supplier No. 05401
MesenCult™ MSC Basal Medium (Human) is a standardized basal medium designed to be supplemented with MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human; Catalog #05402) for the in vitro culture of human mesenchymal stem cells (MSCs). MesenCult™ MSC Basal Medium is a component of MesenCult™ Proliferation Kit (Human; Catalog #05411), and is also available separately.