Learn More
STEMCELL Technologies MesenCult™ MSC Stimulatory Supplement (Human), Size: 50 mL
SDP
Catalog No. 501974345
Encompass
MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is a standardized, serum-containing supplement for the culture of human mesenchymal stem cells (MSCs). MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is optimized for the expansion of human mesenchymal stem cells in vitro as well as their enumeration using the colony-forming unit - fibroblast (CFU-F) assay when combined with MesenCult™ MSC Basal Medium (Human). Components are pre-screened to minimize lot-to-lot variability. This supplement is also a component of MesenCult™ Proliferation Kit (Human; Catalog #05411).
Catalog No. 50-197-4345 Supplier STEMCELL Technologies Supplier No. 05402
MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is a standardized, serum-containing supplement for the culture of human mesenchymal stem cells (MSCs). MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is optimized for the expansion of human mesenchymal stem cells in vitro as well as their enumeration using the colony-forming unit - fibroblast (CFU-F) assay when combined with MesenCult™ MSC Basal Medium (Human). Components are pre-screened to minimize lot-to-lot variability. This supplement is also a component of MesenCult™ Proliferation Kit (Human; Catalog #05411).