Learn More
STEMCELL Technologies MesenCult™ Osteogenic Stimulatory Kit (Mouse), Size: 250 mL
SDP
Catalog No. NC1439695
Encompass
MesenCult™ Osteogenic Stimulatory Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs) from compact bone and bone marrow, and mouse embryonic fibroblasts (MEFs) into osteoblasts. This kit induces strong osteogenesis of mouse MSCs and MEFs, as evidenced by qPCR analysis of key transcripts involved in bone differentiation and maturation, and Alizarin Red staining. This kit is recommended for characterizing MSCs and MEFs and studying bone development.
Catalog No. NC1439695 Supplier STEMCELL Technologies Supplier No. 05504
MesenCult™ Osteogenic Stimulatory Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs) from compact bone and bone marrow, and mouse embryonic fibroblasts (MEFs) into osteoblasts. This kit induces strong osteogenesis of mouse MSCs and MEFs, as evidenced by qPCR analysis of key transcripts involved in bone differentiation and maturation, and Alizarin Red staining. This kit is recommended for characterizing MSCs and MEFs and studying bone development.