Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Mesenpan: xeno-free, serum-free medium for human mesenchymal stem cells (hMSC) expansion (500 ml kit)
SDP
Catalog No. 502388633
Encompass_Preferred
Mesenpan: xeno-free, serum-free medium for human mesenchymal stem cells (hMSC) expansion (500 ml) QUANTITY: 1 kit. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: Media: 2 to 8 degrees Celsius; Supplement: -20 degrees Celsius. DESCRIPTION: Mesenpan Special Medium is a xeno-free, serum-free medium that was developed especially for proliferation of human mesenchymal stem cells (hMSC) without cell differentiation. It allows a long time culture from hMSC while preserving the differentiation potential in different cell types (multi-lineage potential). To reach a differentiation of the expanded hMSC into different lines, the suitable differentiation factors and protocols should be used. COMPOSITION: Media contains salts, amino acids, trace elements, hormones and growth factors in an optimized formulation. Kit includes 500 ml Mesenpan Basal Medium and 5 ml Mesenpan Growth Supplement.
Catalog No. 50-238-8633 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0850400K
Mesenpan: xeno-free, serum-free medium for human mesenchymal stem cells (hMSC) expansion (500 ml) QUANTITY: 1 kit. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: Media: 2 to 8 degrees Celsius; Supplement: -20 degrees Celsius. DESCRIPTION: Mesenpan Special Medium is a xeno-free, serum-free medium that was developed especially for proliferation of human mesenchymal stem cells (hMSC) without cell differentiation. It allows a long time culture from hMSC while preserving the differentiation potential in different cell types (multi-lineage potential). To reach a differentiation of the expanded hMSC into different lines, the suitable differentiation factors and protocols should be used. COMPOSITION: Media contains salts, amino acids, trace elements, hormones and growth factors in an optimized formulation. Kit includes 500 ml Mesenpan Basal Medium and 5 ml Mesenpan Growth Supplement.