Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ MesenPRO RS™ Medium

Catalog No. 12746012
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-746-012 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-746-012 Supplier Gibco™ Supplier No. 12746012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MesenPRO RS™ Medium

MesenPRO RS™ Medium is a reduced serum (2%) medium specifically formulated to support the growth of human mesenchymal stem cells (MSCs) in culture. MesenPRO RS™ Medium consistently improves MSC expansion compared with conventional serum-supplemented media. Using MesenPRO RS™ Medium, MSCs can be expanded for multiple passages while maintaining their multipotential phenotype (i.e., differentiation into osteogenic, chondrogenic, and adipogenic lineages).

MesenPRO RS™ Medium consists of:
  • Basal liquid medium (MesenPRO RS™ Basal Medium)
  • Frozen supplement (MesenPRO RS™ Growth Supplement), which is added at the time of use.
Applications:

Formulated for the growth and expansion of mesenchymal stem cells.

Note: MesenPRO RS™ Medium has been developed for culturing MSCs at greater than clonal densities.

Recommended storage conditions:

  • MesenPRO RS™ Basal Medium: Store in the dark at 2 to 8°C
  • MesenPRO RS™ Growth Supplement: Store in the dark at -5 to -20°C

Both the Basal Medium and Growth Supplement are guaranteed stable for 24 months when properly stored.

Note: MesenPRO RS™ complete medium (basal, supplement and glutamine) is stable for 15 days when stored in the dark at 2 to 8°C. We do not recommend using the complete medium beyond 15 days.

Intended use(s):

in vitro diagnostic (IVD)

This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesenchymal Stem Cells
Classification Serum-supplemented
Form Liquid
Product Type Stem Cell Media
Serum Level Reduced Serum
Without Additives No Glutamine, No Phenol Red
Product Line MesenPRO
Quantity 1 Kit
Species Human
Content And Storage MesenPRO RS™ Medium consists of:
• 500 ml MesenPRO RS™ Basal Medium (store in the dark at 2°C to 8°C)
• 10 ml MesenPRO RS™ Growth Supplement (store in the dark at -5°C to -20°C)

The Basal Medium ships at room temperature, the Growth Supplement on dry ice.
Culture Type Adherent Cell Culture
Show More Show Less
Can I use MesenPRO RS Medium with both StemPro Human Adipose-Derived Stem Cells and StemPro Bone Marrow (BM) Mesenchymal Stem Cells?

Yes, the MesenPRO RS Medium (with growth supplement added) can be used for both cell types.

What kinds of growth factors are used in mesenchymal stem cell (MSC) research?

Growth factors used for MSC maintenance and self-renewal: LIF, FGF-basic, EGF, HGF, PDGF, and Wnt3.

Growth factors or cytokines for MSC differentiation: BMP-2and TGF-β1.

(Arthritis Res Ther 9:204 (2007)

View our Gibco™ MSC research products.

How can I culture mesenchymal stem cells (MSCs) in vitro?

MSCs may be cultured in either serum-containing medium or serum-free medium. The standard culturing conditions are DMEM, low glucose with 10% FBS. MSCs can also be grown in reduced-serum (2%) Gibco MesenPRO RS Medium (Cat. No. 12746-012), or in serum-free Gibco StemPro MSC SFM XenoFree Medium (Cat. No. A10675) and Gibco StemPro MSC SFM (Cat. No. A10332). In general, MSCs grow better under hypoxic conditions (2% O2).

How do you characterize human mesenchymal stem cells (MSCs)?

Human MSCs can be identified by flow cytometry, typically displaying CD73+, CD90+, CD105+, CD11b-, and CD45- marker characteristics (Blood 109:4245 (2007)). Human MSC expresses Oct4, Sox2, and Rex-1; these may be verified using RT-PCR (Arthritis Res Ther 9:204 (2007)).

What are mesenchymal stem cells?

Mesenchymal stem cells (MSCs) are multipotent cells isolated primarily from bone marrow or fat tissues that exhibit the ability to differentiate into bone, cartilage, and fat cells. Under normal cell culture conditions, MSCs isolated from bone marrow are spindle shaped with the unique ability to adhere to uncoated plastic culture dishes (Arthritis Res Ther 9:204 (2007)).

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.