Learn More
MSC PIP Back Support Belt, Nylon Mesh, 9 in., XL, 1 ea.
Catalog No. 5000687290
Encompass
This 9 in. back support belt is designed for use in heavy-duty work environments to provide lumbar stability and help reduce the risk of back injuries. Constructed with breathable nylon mesh, the belt provides consistent support and a secure, adjustable fit for daily wear.
  • Breathable nylon mesh body
  • Five back stays to prevent belt from riding up
  • Front panels with hook and loop closure for a secure, custom fit
  • Detachable and adjustable elastic suspenders
  • Belt width of 9 in.
  • Fits waist sizes 44 in. to 48 in.
Catalog No. 50-006-87290 Supplier MSC Supplier No. 51285328
This 9 in. back support belt is designed for use in heavy-duty work environments to provide lumbar stability and help reduce the risk of back injuries. Constructed with breathable nylon mesh, the belt provides consistent support and a secure, adjustable fit for daily wear.
  • Breathable nylon mesh body
  • Five back stays to prevent belt from riding up
  • Front panels with hook and loop closure for a secure, custom fit
  • Detachable and adjustable elastic suspenders
  • Belt width of 9 in.
  • Fits waist sizes 44 in. to 48 in.