Learn More
Marlin Steel Wire Products Round Mesh Basket with Handles 12"D x 8"H
SDP
Catalog No. NC1962228
Encompass_Preferred
Round material handling basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Stackable Basket4 mesh openings per linear inch"
Catalog No. NC1962228 Supplier Marlin Steel Wire Products Supplier No. 0010131
Add to Cart
Add to Cart
Round material handling basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Stackable Basket4 mesh openings per linear inch"