Learn More
Marlin Steel Wire Products Mesh Basket with Handles 12"L x 18.12"W x 9"H
SDP
Catalog No. NC1962234
Encompass_Preferred
Rectangular material handling basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Nestable Basket4 mesh openings per linear inch
Catalog No. NC1962234 Supplier Marlin Steel Wire Products Supplier No. 00105A31
Rectangular material handling basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Nestable Basket4 mesh openings per linear inch